siRNA Details
| Gene Symbol: Terc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_001567.1 | LIT2120 telomerase RNA |
1 | 55 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF552926.1 | - | 47 | Chr. - - [-] on Build | ||

Terc
Genomic Map