siRNA Details
| Gene Symbol: Rmrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002703.1 | 1 | 245 | |||
| GenBank mRNA | L08800.1 | - | 1 | 273 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rmrp| Gene Symbol: Rmrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002703.1 | 1 | 245 | |||
| GenBank mRNA | L08800.1 | - | 1 | 273 | ||