siRNA Details
| Gene Symbol: Dcm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033150.1 | 1 | 548 | |||
| GenBank mRNA | AF412817.1 | - | 1 | 549 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcm5| Gene Symbol: Dcm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033150.1 | 1 | 548 | |||
| GenBank mRNA | AF412817.1 | - | 1 | 549 | ||