siRNA Details
| Gene Symbol: RGD1559747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027235.1 | 4 | 1403 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK600316.1 | - | 320 | Chr. - - [-] on Build | ||

RGD1559747
Genomic Map