siRNA Details
| Gene Symbol: Cb707485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029398.1 | 2, 3 | 219 | |||
| GenBank mRNA | CB707485.1 | - | 2, 3 | 232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cb707485| Gene Symbol: Cb707485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029398.1 | 2, 3 | 219 | |||
| GenBank mRNA | CB707485.1 | - | 2, 3 | 232 | ||