siRNA Details
| Gene Symbol: 4933431E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015459.1 | 3 | 5229 | |||
| GenBank mRNA | AK134641.1 | - | 1 | 2312 | ||
| BC060136.1 | - | 3 | 5229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933431E20Rik| Gene Symbol: 4933431E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015459.1 | 3 | 5229 | |||
| GenBank mRNA | AK134641.1 | - | 1 | 2312 | ||
| BC060136.1 | - | 3 | 5229 | |||