siRNA Details
| Gene Symbol: Snora43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028572.1 | 1 | 81 | |||
| Gene Symbol: Snhg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC046478.1 | - | 3 | 1732 | ||

Snora43
Genomic Map