siRNA Details
| Gene Symbol: Tmem51os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027137.1 | 4, 5 | 1400 | |||
| GenBank mRNA | AK154034.1 | - | 4, 5 | 1358 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY763106.1 | - | 1, 2 | 398 | Chr. 4 - 142086892 - 142088045 [+] on Build GRCm38 | |

Tmem51os1
Genomic Map