siRNA Details
Gene Symbol: Tmem51os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027137.1 | 3 | 899 | |||
GenBank mRNA | AK153738.1 | - | 2 | 1850 | ||
AK154034.1 | - | 3 | 857 | |||
AK158244.1 | - | 3 | 817 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem51os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027137.1 | 3 | 899 | |||
GenBank mRNA | AK153738.1 | - | 2 | 1850 | ||
AK154034.1 | - | 3 | 857 | |||
AK158244.1 | - | 3 | 817 |