siRNA Details
Gene Symbol: BC025920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030677.1 | 1 | 52 | |||
GenBank mRNA | AK140772.1 | - | 1 | 53 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC025920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030677.1 | 1 | 52 | |||
GenBank mRNA | AK140772.1 | - | 1 | 53 |