siRNA Details
| Gene Symbol: Gm7854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028417.1 | 2 | 486 | |||
| GenBank mRNA | AK041797.1 | - | 2 | 487 | ||
| AK138349.1 | - | 2 | 117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7854| Gene Symbol: Gm7854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028417.1 | 2 | 486 | |||
| GenBank mRNA | AK041797.1 | - | 2 | 487 | ||
| AK138349.1 | - | 2 | 117 | |||