siRNA Details
| Gene Symbol: Mir710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030491.1 | 1 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mir710| Gene Symbol: Mir710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030491.1 | 1 | 74 | |||