siRNA Details
| Gene Symbol: Gm16386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030709.2 | 5 | 537 | |||
| GenBank mRNA | BG086466.2 | - | 5 | 537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16386| Gene Symbol: Gm16386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030709.2 | 5 | 537 | |||
| GenBank mRNA | BG086466.2 | - | 5 | 537 | ||