siRNA Details
| Gene Symbol: Gm16386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030709.2 | 1 | 58 | |||
| GenBank mRNA | AK144317.1 | - | 1 | 52 | ||
| BG086466.2 | - | 1 | 58 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16386| Gene Symbol: Gm16386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030709.2 | 1 | 58 | |||
| GenBank mRNA | AK144317.1 | - | 1 | 52 | ||
| BG086466.2 | - | 1 | 58 | |||