siRNA Details
| Gene Symbol: Mir697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030479.1 | 1 | 33 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB057993.1 | - | 2 | 366 | Chr. 4 - 124730585 - 124732014 [+] on Build GRCm38 | |

Mir697
Genomic Map