siRNA Details
| Gene Symbol: Atcayos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015477.1 | 3 | 262 | |||
| GenBank mRNA | AK033837.1 | - | 489 | |||
| AK084504.1 | - | 3 | 262 | |||
| AK136697.1 | - | 495 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atcayos| Gene Symbol: Atcayos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015477.1 | 3 | 262 | |||
| GenBank mRNA | AK033837.1 | - | 489 | |||
| AK084504.1 | - | 3 | 262 | |||
| AK136697.1 | - | 495 | ||||