siRNA Details
| Gene Symbol: Cdk3-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004853.1 | 7 | 1040 | |||
| GenBank mRNA | AK009918.1 | - | 5 | 866 | ||
| AK166387.1 | - | 2 | 353 | |||
| BC116895.1 | - | 5 | 776 | |||
| BC119034.1 | - | 5 | 776 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM934713.1 | - | 4 | 569 | Chr. 11 - 116216928 - 116219934 [+] on Build GRCm38 | |

Cdk3-ps
Genomic Map