siRNA Details
| Gene Symbol: Cdk3-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004853.1 | 7 | 1246 | |||
| GenBank mRNA | AK009918.1 | - | 5 | 1072 | ||
| AK166387.1 | - | 2 | 559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk3-ps| Gene Symbol: Cdk3-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004853.1 | 7 | 1246 | |||
| GenBank mRNA | AK009918.1 | - | 5 | 1072 | ||
| AK166387.1 | - | 2 | 559 | |||