siRNA Details
| Gene Symbol: 4930470P17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027825.1 | 1 | 728 | |||
| GenBank mRNA | AK015542.1 | - | 1 | 730 | ||
| AK015810.1 | - | 1 | 728 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930470P17Rik| Gene Symbol: 4930470P17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027825.1 | 1 | 728 | |||
| GenBank mRNA | AK015542.1 | - | 1 | 730 | ||
| AK015810.1 | - | 1 | 728 | |||