siRNA Details
| Gene Symbol: 4930483J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015603.1 | 2 | 653 | |||
| GenBank mRNA | AK015615.1 | - | 2 | 533 | ||
| AK016854.1 | - | 2 | 654 | |||
| BC049560.1 | - | 4 | 576 | |||
| BC087915.1 | - | 1 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930483J18Rik| Gene Symbol: 4930483J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015603.1 | 2 | 653 | |||
| GenBank mRNA | AK015615.1 | - | 2 | 533 | ||
| AK016854.1 | - | 2 | 654 | |||
| BC049560.1 | - | 4 | 576 | |||
| BC087915.1 | - | 1 | 251 | |||