siRNA Details
Gene Symbol: 4930483J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015603.1 | 2 | 202 | |||
GenBank mRNA | AK015615.1 | - | 2 | 82 | ||
AK016854.1 | - | 2 | 203 | |||
BC049560.1 | - | 3 | 423 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930483J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015603.1 | 2 | 202 | |||
GenBank mRNA | AK015615.1 | - | 2 | 82 | ||
AK016854.1 | - | 2 | 203 | |||
BC049560.1 | - | 3 | 423 |