siRNA Details
| Gene Symbol: 4930483J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015603.1 | 2 | 393 | |||
| GenBank mRNA | AK015615.1 | - | 2 | 273 | ||
| AK016854.1 | - | 2 | 394 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716529.1 | - | 2 | 394 | Chr. 15 - 81190853 - 81192603 [+] on Build GRCm38 | |

4930483J18Rik
Genomic Map