siRNA Details
| Gene Symbol: 4930483J18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015603.1 | 2 | 941 | |||
| GenBank mRNA | AK015615.1 | - | 2 | 821 | ||
| AK016854.1 | - | 2 | 942 | |||
| AV322473.1 | - | 1 | 109 | |||
| BC049560.1 | - | 4 | 864 | |||
| BC087915.1 | - | 1 | 539 | |||

4930483J18Rik
Genomic Map