siRNA Details
| Gene Symbol: 4930465K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027978.1 | 1 | 984 | |||
| GenBank mRNA | AK015507.1 | - | 1 | 957 | ||
| BC061021.1 | - | 1 | 964 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930465K10Rik| Gene Symbol: 4930465K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027978.1 | 1 | 984 | |||
| GenBank mRNA | AK015507.1 | - | 1 | 957 | ||
| BC061021.1 | - | 1 | 964 | |||