siRNA Details
| Gene Symbol: 4930515G01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027872.1 | 1 | 563 | |||
| GenBank mRNA | AK015794.1 | - | 1 | 177 | ||
| AK021339.1 | - | 1 | 166 | |||
| AK041663.1 | - | 1 | 564 | |||
| BC147457.1 | - | 1 | 507 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930515G01Rik| Gene Symbol: 4930515G01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027872.1 | 1 | 563 | |||
| GenBank mRNA | AK015794.1 | - | 1 | 177 | ||
| AK021339.1 | - | 1 | 166 | |||
| AK041663.1 | - | 1 | 564 | |||
| BC147457.1 | - | 1 | 507 | |||