siRNA Details
Gene Symbol: Vmn2r-ps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004441.1 | 7 | 2034 | |||
GenBank mRNA | AK030272.1 | - | 7 | 2035 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn2r-ps54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004441.1 | 7 | 2034 | |||
GenBank mRNA | AK030272.1 | - | 7 | 2035 |