siRNA Details
| Gene Symbol: 4930523C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028111.1 | 3 | 888 | |||
| GenBank mRNA | AK015875.1 | - | 3 | 855 | ||
| AK041441.1 | - | 3 | 684 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930523C07Rik| Gene Symbol: 4930523C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028111.1 | 3 | 888 | |||
| GenBank mRNA | AK015875.1 | - | 3 | 855 | ||
| AK041441.1 | - | 3 | 684 | |||