siRNA Details
| Gene Symbol: 4930558C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015490.1 | 2 | 541 | |||
| GenBank mRNA | AK006075.1 | - | 2 | 486 | ||
| AK006566.1 | - | 2 | 508 | |||
| AK016169.1 | - | 2 | 532 | |||
| BC049745.1 | - | 2 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930558C23Rik| Gene Symbol: 4930558C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015490.1 | 2 | 541 | |||
| GenBank mRNA | AK006075.1 | - | 2 | 486 | ||
| AK006566.1 | - | 2 | 508 | |||
| AK016169.1 | - | 2 | 532 | |||
| BC049745.1 | - | 2 | 541 | |||