siRNA Details
| Gene Symbol: 4930592I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033307.1 | 3 | 915 | |||
| GenBank mRNA | AK077183.1 | - | 3 | 904 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930592I03Rik| Gene Symbol: 4930592I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033307.1 | 3 | 915 | |||
| GenBank mRNA | AK077183.1 | - | 3 | 904 | ||