siRNA Details
| Gene Symbol: Fate1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003243.2 | 1 | 31 | |||
| GenBank mRNA | AK020359.1 | - | 1 | 32 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721172.1 | - | 1 | 30 | Chr. X - 71972987 - 71988731 [+] on Build GRCm38 | |

Fate1
Genomic Map