siRNA Details
| Gene Symbol: Speer4cos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001585.3 | 5 | 1035 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK016255.1 | - | 1034 | Chr. - - [-] on Build | ||

Speer4cos
Genomic Map