siRNA Details
| Gene Symbol: 0610040B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027874.1 | 2 | 407 | |||
| GenBank mRNA | AK002861.1 | - | 2 | 408 | ||
| AK009603.1 | - | 2 | 409 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703359.1 | - | 2 | 408 | Chr. 5 - 143329308 - 143332696 [+] on Build GRCm38 | |

0610040B10Rik
Genomic Map