siRNA Details
| Gene Symbol: 0610040B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027874.1 | 2 | 168 | |||
| GenBank mRNA | AK002861.1 | - | 2 | 169 | ||
| AK009603.1 | - | 2 | 170 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

0610040B10Rik| Gene Symbol: 0610040B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027874.1 | 2 | 168 | |||
| GenBank mRNA | AK002861.1 | - | 2 | 169 | ||
| AK009603.1 | - | 2 | 170 | |||