siRNA Details
| Gene Symbol: Gm17801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027452.1 | 1 | 1138 | |||
| GenBank mRNA | AK143641.1 | - | 1 | 1139 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB735807.1 | - | 1 | 139 | Chr. 17 - 25275123 - 25275528 [+] on Build GRCm38 | |

Gm17801
Genomic Map