siRNA Details
| Gene Symbol: Gm17801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027452.1 | 1 | 733 | |||
| GenBank mRNA | AK143641.1 | - | 1 | 733 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17801| Gene Symbol: Gm17801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027452.1 | 1 | 733 | |||
| GenBank mRNA | AK143641.1 | - | 1 | 733 | ||