siRNA Details
| Gene Symbol: 2410018L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015504.1 | 4 | 1101 | |||
| GenBank mRNA | AK134640.1 | - | 4 | 1102 | ||
| BC063063.1 | - | 5 | 3071 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410018L13Rik| Gene Symbol: 2410018L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015504.1 | 4 | 1101 | |||
| GenBank mRNA | AK134640.1 | - | 4 | 1102 | ||
| BC063063.1 | - | 5 | 3071 | |||