siRNA Details
| Gene Symbol: 2410018L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015504.1 | 2, 3 | 458 | |||
| GenBank mRNA | AK134640.1 | - | 2, 3 | 459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410018L13Rik| Gene Symbol: 2410018L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015504.1 | 2, 3 | 458 | |||
| GenBank mRNA | AK134640.1 | - | 2, 3 | 459 | ||