siRNA Details
| Gene Symbol: Gm5607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027975.2 | 5 | 1148 | |||
| GenBank mRNA | BC066076.1 | - | 5 | 1148 | 6852276 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5607| Gene Symbol: Gm5607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027975.2 | 5 | 1148 | |||
| GenBank mRNA | BC066076.1 | - | 5 | 1148 | 6852276 | |