siRNA Details
| Gene Symbol: A930004D18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028376.1 | 1 | 108 | |||
| NR_028377.1 | 1 | 108 | ||||
| GenBank mRNA | AK020807.1 | - | 1 | 112 | ||
| AK044329.1 | - | 1 | 105 | |||
| AK044517.1 | - | 1 | 90 | |||
| EL956282.1 | - | 1 | 112 | |||

A930004D18Rik
Genomic Map