siRNA Details
| Gene Symbol: Gm8801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028278.1 | 5 | 655 | |||
| GenBank mRNA | CF916400.1 | - | 5 | 598 | ||
| DV649894.1 | - | 5 | 633 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8801| Gene Symbol: Gm8801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028278.1 | 5 | 655 | |||
| GenBank mRNA | CF916400.1 | - | 5 | 598 | ||
| DV649894.1 | - | 5 | 633 | |||