siRNA Details
| Gene Symbol: Gm8801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028278.1 | 2 | 401 | |||
| GenBank mRNA | CA558512.1 | - | 2 | 401 | ||
| CF916400.1 | - | 2 | 344 | |||
| DV649894.1 | - | 2 | 379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8801| Gene Symbol: Gm8801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028278.1 | 2 | 401 | |||
| GenBank mRNA | CA558512.1 | - | 2 | 401 | ||
| CF916400.1 | - | 2 | 344 | |||
| DV649894.1 | - | 2 | 379 | |||