siRNA Details
Gene Symbol: Gm8801 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028278.1 | 5, 6 | 739 | |||
GenBank mRNA | DV649894.1 | - | 5, 6 | 718 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm8801 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028278.1 | 5, 6 | 739 | |||
GenBank mRNA | DV649894.1 | - | 5, 6 | 718 |