siRNA Details
| Gene Symbol: Gas5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002840.2 | 1 | 1439 | |||
| GenBank mRNA | AK083453.1 | - | 1 | 1440 | ||
| BC080708.1 | - | 2 | 686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gas5| Gene Symbol: Gas5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002840.2 | 1 | 1439 | |||
| GenBank mRNA | AK083453.1 | - | 1 | 1440 | ||
| BC080708.1 | - | 2 | 686 | |||