siRNA Details
| Gene Symbol: Gas5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002840.2 | 1 | 696 | |||
| GenBank mRNA | AK083453.1 | - | 1 | 697 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF608434.1 | - | 137 | Chr. - - [-] on Build | ||

Gas5
Genomic Map