siRNA Details
| Gene Symbol: 1810058I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015608.1 | 3 | 672 | |||
| NR_027875.1 | 3 | 902 | ||||
| GenBank mRNA | AK007895.1 | - | 3 | 662 | ||
| AK082166.1 | - | 718 | ||||
| AK155176.1 | - | 3 | 662 | |||
| AK158055.1 | - | 3 | 903 | |||
| CA751247.1 | - | 1 | 235 | |||

1810058I24Rik
Genomic Map