siRNA Details
| Gene Symbol: 1810058I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015608.1 | 3 | 996 | |||
| NR_027875.1 | 3 | 1226 | ||||
| GenBank mRNA | AK082166.1 | - | 1042 | |||
| CA751247.1 | - | 1 | 559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810058I24Rik| Gene Symbol: 1810058I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015608.1 | 3 | 996 | |||
| NR_027875.1 | 3 | 1226 | ||||
| GenBank mRNA | AK082166.1 | - | 1042 | |||
| CA751247.1 | - | 1 | 559 | |||