siRNA Details
| Gene Symbol: 4932414J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028259.1 | 7 | 2478 | |||
| GenBank mRNA | AK030001.1 | - | 7 | 2479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4932414J04Rik| Gene Symbol: 4932414J04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028259.1 | 7 | 2478 | |||
| GenBank mRNA | AK030001.1 | - | 7 | 2479 | ||