siRNA Details
Gene Symbol: Olfr75-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002859.2 | 2 | 897 | |||
NR_033528.1 | 3 | 1116 | ||||
GenBank mRNA | Y15524.1 | - | 1 | 595 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr75-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002859.2 | 2 | 897 | |||
NR_033528.1 | 3 | 1116 | ||||
GenBank mRNA | Y15524.1 | - | 1 | 595 |