siRNA Details
| Gene Symbol: Cep83os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015524.1 | 3 | 3797 | |||
| GenBank mRNA | AK016525.1 | - | 3 | 3757 | ||
| AK034425.1 | - | 308 | ||||
| AK144701.1 | - | 3 | 3761 | |||
| AK154718.1 | - | 3 | 3730 | |||
| AK155868.1 | - | 3 | 3730 | |||
| BB626469.1 | - | 1 | 308 | |||
| BC021358.1 | - | 1 | 2576 | |||
| BC061482.1 | - | 1 | 2773 | |||
| BC067070.1 | - | 3 | 3743 | |||

Cep83os
Genomic Map