siRNA Details
| Gene Symbol: Cep83os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015524.1 | 3 | 955 | |||
| GenBank mRNA | AK015136.1 | - | 3 | 869 | ||
| AK016525.1 | - | 3 | 914 | |||
| AK085032.1 | - | 3 | 921 | |||
| AK144701.1 | - | 3 | 917 | |||
| AK154718.1 | - | 3 | 887 | |||
| AK155868.1 | - | 3 | 887 | |||
| BC067070.1 | - | 3 | 900 | |||

Cep83os
Genomic Map